Etidronate disodium
Etidronate disodium
Etidronate disodium
Thermo Scientific Chemicals

Etidronate disodium

CAS: 7414-83-7 | C2H6Na2O7P2 | 249.99 g/mol
Have Questions?
Catalog NumberQuantity
J62381.03
also known as J62381-03
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62381.03
also known as J62381-03
Price (USD)
243.65
Special offer
Online exclusive
Ends: 28-Dec-2026
286.00
Save 42.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
243.65
Special offer
Online exclusive
Ends: 28-Dec-2026
286.00
Save 42.35
Each
Specifications
CAS7414-83-7
ChEBICHEBI:4906
Chemical Name or MaterialEtidronate disodium
ColorWhite
EINECS Number231-025-7
View more
Etidronate disodium is used as a bisphosphonate bone resorption inhibitor. Etidronic acid is used as a calcium regulator. Etidronic acid is a bisphosphonate antiresorptive agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Etidronate disodium is used as a bisphosphonate bone resorption inhibitor. Etidronic acid is used as a calcium regulator. Etidronic acid is a bisphosphonate antiresorptive agent.

Solubility
Soluble in water (26 mg/m
RUO – Research Use Only

General References:

  1. RP Heaney.; PD Saville. Etidronate disodium in postmenopausal osteoporosis. Clinical pharmacology and therapeutics. 1976, 20(5), 593-604.
  2. N Shiraishi.; K Kitamura.;T Miyoshi.; M Adachi. Successful treatment of a patient with severe calcific uremic arteriolopathy (calciphylaxis) byetidronate disodium. American Journal of Kidney Diseases. 2006, 48(1), 151-154.