Tetraethylene glycol monooctyl ether
Tetraethylene glycol monooctyl ether
Tetraethylene glycol monooctyl ether
Thermo Scientific Chemicals

Tetraethylene glycol monooctyl ether

Reagent for solubilizing membrane proteins | CAS: 19327-39-0 | C16H34O5 | 306.443 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J62496.03
also known as J62496-03
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Specifications
Beilstein1781226
CAS19327-39-0
ChEBICHEBI:41323
Chemical Name or MaterialTetraethylene glycol monooctyl ether
ColorColorless
View more
Reagent for solubilizing membrane proteins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only