MES, 0.5M buffer soln., pH 5.5 - FAQs
MES, 0.5M buffer soln., pH 5.5 - FAQs
View additional product information for MES, 0.5M buffer soln., pH 5.5 - FAQs (J62534.AE, J62534.AK)
1 product FAQs found
What is the density of MES, 0.5M buffer soln., pH 5.5 (Cat. No. J62534.AK)?
The density of MES, 0.5 M buffer soln., pH 5.5 is 1.07 g/cm3.