Magnesium chloride hexahydrate, Cell Culture Reagent
Magnesium chloride hexahydrate, Cell Culture Reagent
Magnesium chloride hexahydrate, Cell Culture Reagent
Magnesium chloride hexahydrate, Cell Culture Reagent
Thermo Scientific Chemicals

Magnesium chloride hexahydrate, Cell Culture Reagent

CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
Catalog number J62575.36
also known as J62575-36
Price (USD)
78.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.80
Save 13.15 (14%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
78.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.80
Save 13.15 (14%)
Each
Add to cart
Chemical Identifiers
CAS7791-18-6
IUPAC Namemagnesium(2+) hexahydrate dichloride
Molecular FormulaCl2H12MgO6
InChI KeyDHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSulfate: ≤0.002%
Heavy Metals≤5 ppm (as Pb)
Assay from Supplier's CofA≥99.0% (titration)
CommentAmmonium: ≤0.002%
CommentBarium: ≤0.005%
View more
Magnesium Chloride, Hexahydrate is widely used as a source of magnesium ion in chemistry and molecular biology applications. In biological systems, magnesium is a co-factor for many enzymes including deoxyribonuclease (DNase) and various restriction enzymes. Also plays a role in cell membrane integrity, muscle cell physiology, cardiovascular and muscular activity, and nucleic acid structure. Magnesium chloride solution is a favorable choice as an elution buffer for antibody affinity column purifications; it is much milder on most antigens than acid elution, allowing reuse of the antigen column. Also an essential cofactor for the DNA polymerase in polymerase chain reaction (PCR) amplification. Used in wood fireproofing, wool carbonizing, and cotton fabric dressing; Also used in disinfectants, magnesia cements, fire extinguishers, floor sweeping compounds, and casein glue; Also used to make parchment paper and artificial leather; Has been used therapeutically as a cathartic and to treat bovine hypomagnesaemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium Chloride, Hexahydrate is widely used as a source of magnesium ion in chemistry and molecular biology applications. In biological systems, magnesium is a co-factor for many enzymes including deoxyribonuclease (DNase) and various restriction enzymes. Also plays a role in cell membrane integrity, muscle cell physiology, cardiovascular and muscular activity, and nucleic acid structure. Magnesium chloride solution is a favorable choice as an elution buffer for antibody affinity column purifications; it is much milder on most antigens than acid elution, allowing reuse of the antigen column. Also an essential cofactor for the DNA polymerase in polymerase chain reaction (PCR) amplification. Used in wood fireproofing, wool carbonizing, and cotton fabric dressing; Also used in disinfectants, magnesia cements, fire extinguishers, floor sweeping compounds, and casein glue; Also used to make parchment paper and artificial leather; Has been used therapeutically as a cathartic and to treat bovine hypomagnesaemia.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Brooks JE. Properties and uses of restriction endonucleases.Methods Enzymol.,1987,152113-29.
  2. Huang HW, et al. Metallobiochemistry of the magnesium ion. Characterization of the essential metal-binding site in Escherichia coli ribonuclease H.Eur J Biochem.,1994,219(1-2), 253-60.