Diclofenac sodium salt
Diclofenac sodium salt
Diclofenac sodium salt
Thermo Scientific Chemicals

Diclofenac sodium salt

Standard NSAID and cyclooxygenase (COX) inhibitor | CAS: 15307-79-6 | C14H10Cl2NNaO2 | 318.129 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62609.14
also known as J62609-14
Price (USD)
80.65
Special offer
Online exclusive
Ends: 30-Jun-2026
94.20
Save 13.55 (14%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
80.65
Special offer
Online exclusive
Ends: 30-Jun-2026
94.20
Save 13.55 (14%)
Each
Chemical Identifiers
CAS15307-79-6
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay from Supplier's CofA≥97.5%
Oxidation of diclofenac sodium produces the metabolite 4'-hydroxy diclofenac) which demonstrates specific inhibition of Cox-2. Inhibition of Cox by diclofenac and 4'-hydroxy diclofenac suppresses prostaglandin E2 synthesis, producing anti-inflammatory and analgesic effects. Diclofenac is also shown to stabilize the native tetrameric conformation of transthyretin (TTR) fibrils, preventing the formation of insoluble amyloidogenic TTR deposits. Diclofenac Sodium is a substrate of CYP2C9. It is also used as an inhibitor of Cox-1 and Cox-2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxidation of diclofenac sodium produces the metabolite 4′-hydroxy diclofenac) which demonstrates specific inhibition of Cox-2. Inhibition of Cox by diclofenac and 4′-hydroxy diclofenac suppresses prostaglandin E2 synthesis, producing anti-inflammatory and analgesic effects. Diclofenac is also shown to stabilize the native tetrameric conformation of transthyretin (TTR) fibrils, preventing the formation of insoluble amyloidogenic TTR deposits. Diclofenac Sodium is a substrate of CYP2C9. It is also used as an inhibitor of Cox-1 and Cox-2.

Solubility
Soluble in water (50 mg/ml), PBS pH 7.2 (6 mg/ml), ethanol (∼35 mg/ml), DMF (∼35 mg/ml), DMSO (∼35 mg/ml), methanol (>24 mg/ml), and acetone (6 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Barnett, J., et al. Purification, characterization and selective inhibition of human prostaglandin G/H synthase 1 and 2 expressed in the baculovirus system. Biochim. Biophys. Acta. 1994, 1209, (1), 130-9.
  2. Laneuville, O., et al. Differential inhibition of human prostaglandin endoperoxide H synthases-1 and -2 by nonsteroidal anti-inflammatory drugs. J. Pharmacol. Exp. Ther. 1994, 271, (2), 927-34.