Stains-All, 95%
Stains-All, 95%
Stains-All, 95%
Thermo Scientific Chemicals

Stains-All, 95%

Cationic carbocyanine dye used in electrophoresis to stain proteins and nucleic acids | CAS: 7423-31-6 | C30H27BrN2S2, C30H27BrN2S2 | 559.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1g
5g
Catalog number J62630.06
also known as J62630-06
Price (USD)
263.65
Online Exclusive
293.00
Save 29.35 (10%)
Each
Quantity:
5g
Request bulk or custom format
Price (USD)
263.65
Online Exclusive
293.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS7423-31-6
IUPAC Name1-ethyl-2-{3-[(2Z)-1-ethyl-1H,2H-naphtho[1,2-d][1,3]thiazol-2-ylidene]-2-methylprop-1-en-1-yl}naphtho[1,2-d][1,3]thiazol-1-ium bromide
Molecular FormulaC30H27BrN2S2
InChI KeyMPBRYMWMMKKRGC-UHFFFAOYSA-M
SMILES[Br-].CCN1\C(SC2=CC=C3C=CC=CC3=C12)=C\C(C)=CC1=[N+](CC)C2=C(S1)C=CC1=CC=CC=C21
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue to dark blue or dark grey or purple
FormCrystals or powder or crystalline powder
Assay from Suppliers CofA≥94.0%
Elemental AnalysisSulfur, S: ≥11.0 to ≤11.46%
lambda-max (UV-Vis)595-601nm
View more
Stains-all is an ideal dye for the visualization and identification of proteins on polyacrylamide gels because it differentiates between highly acidic proteins which stain blue and less acidic proteins which stain pink. When employed on polyacrylamide gels, the intensity of Stains-all can be increased by the addition of silver nitrate. It is a useful nucleic acid and protein stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only