Pyridoxamine dihydrochloride, Cell Culture Reagent
Pyridoxamine dihydrochloride, Cell Culture Reagent
Pyridoxamine dihydrochloride, Cell Culture Reagent
Pyridoxamine dihydrochloride, Cell Culture Reagent
Thermo Scientific Chemicals

Pyridoxamine dihydrochloride, Cell Culture Reagent

CAS: 524-36-7 | C8H14Cl2N2O2 | 241.112 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
5 g
10 g
Catalog number J62679.27
also known as J62679-27
Price (USD)
100.65
Special offer
Online exclusive
Ends: 31-Dec-2025
118.00
Save 17.35 (15%)
Each
Add to cart
Quantity:
2.5 g
Request bulk or custom format
Price (USD)
100.65
Special offer
Online exclusive
Ends: 31-Dec-2025
118.00
Save 17.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS524-36-7
IUPAC Namedihydrogen 4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol dichloride
Molecular FormulaC8H14Cl2N2O2
InChI KeyHNWCOANXZNKMLR-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].CC1=NC=C(CO)C(CN)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Assay from Suppliers CofA≥98.0%
Infrared spectrumConforms
FormPowder or crystals or crystalline powder
Water Content (Karl Fischer Titration)≤10.0%
A vitamin in the Vitamin B-6 family

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only