Phosphate-buffered saline (PBS, 10X), pH 7.6
Thermo Scientific Chemicals

Phosphate-buffered saline (PBS, 10X), pH 7.6

Helps to maintain a constant pH
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2 L
4 L
Catalog number J62692.K3
also known as J62692-K3
Price (USD)
123.00
Each
Quantity:
2 L
Request bulk or custom format
Price (USD)
123.00
Each
Specifications
Chemical Name or MaterialPhosphate Buffered Saline (PBS)
Recommended StorageAmbient temperatures
Concentration10 X
Physical FormLiquid
Quantity2 L
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only