Ketoprofen
Ketoprofen
Ketoprofen
Thermo Scientific Chemicals

Ketoprofen

A non-steroidal anti-inflammatory drug (NSAID); also inhibits cyclooxygenases-1 and -2 | CAS: 22071-15-4 | C16H14O3 | 254.285 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62702.06
also known as J62702-06
Price (USD)
98.90
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
98.90
Each
Chemical Identifiers
CAS22071-15-4
IUPAC Name2-(3-benzoylphenyl)propanoic acid
Molecular FormulaC16H14O3
InChI KeyDKYWVDODHFEZIM-UHFFFAOYNA-N
SMILESCC(C(O)=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0% min.
Melting Point92.0- 97.0?C
It is used as a anti-inflammatory; analgesic and an antipyretic drug cause the hypothalamus to override an interleukin-induced increase in temperature. They act by blocking the synthesis of prostaglandins by inhibiting cyclooxygenase, which converts arachidonic acid to cyclic endoperoxides, precursors of prostaglandins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a anti-inflammatory; analgesic and an antipyretic drug cause the hypothalamus to override an interleukin-induced increase in temperature. They act by blocking the synthesis of prostaglandins by inhibiting cyclooxygenase, which converts arachidonic acid to cyclic endoperoxides, precursors of prostaglandins.

Solubility
Slightly soluble in chloroform and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Osipova NA, T.D, et al. Ketoprofen (ketonal): a drug for preventing and treating postoperative pain.Anesteziologiia i reanimatologiia.,1999,671-74.
  2. Ting ST, et al. Effect of ketoprofen, lidocaine local anesthesia, and combined xylazine and lidocaine caudal epidural anesthesia during castration of beef cattle on stress responses, immunity, growth, and behavior.Journal of animal science.,2003,81(5), 1281-1293.