Mefenamic acid, 98%
Mefenamic acid, 98%
Mefenamic acid, 98%
Thermo Scientific Chemicals

Mefenamic acid, 98%

CAS: 61-68-7 | C15H15NO2 | 241.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62705.14
also known as J62705-14
Price (USD)
39.90
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
39.90
Each
Chemical Identifiers
CAS61-68-7
IUPAC Name2-[(2,3-dimethylphenyl)amino]benzoic acid
Molecular FormulaC15H15NO2
InChI KeyHYYBABOKPJLUIN-UHFFFAOYSA-N
SMILESCC1=CC=CC(NC2=CC=CC=C2C(O)=O)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥97.5% (titration)
Mefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.

Solubility
It is soluble in acetone, chloroform, dichloromethane, methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Hixson, L J., et al . Antiproliferative effect of nonsteroidal antiinflammatory drugs against human colon cancer cells. Cancer epidemiology, biomarkers & prevention . American Association for Cancer Research. 2002, 3, (5), 433-8.
  2. H Gögelein.; D Dahlem.; HC Englert.; HJ Lang. Flufenamic acid, mefenamic acid and niflumic acid inhibit single nonselective cation channels in the rat exocrine pancreas. FEBS letters. 1990, 268, (1), 79-82.