TRIS-buffered saline (TBS, 10X) pH 8.0 - FAQs
TRIS-buffered saline (TBS, 10X) pH 8.0 - FAQs
View additional product information for TRIS-buffered saline (TBS, 10X) pH 8.0 - FAQs (J62780.K7, J62780.K3, J62780.K2)
1 product FAQs found
What are the concentrations of the components of TRIS-buffered saline (TBS, 10X) pH 8.0?
The TRIS-buffered saline (TBS, 10X) pH 8.0 contains 250 mM Tris, 27 mM KCl, and 1.37 M NaCl.