TRIS-buffered saline (TBS, 10X) pH 8.0
TRIS-buffered saline (TBS, 10X) pH 8.0
TRIS-buffered saline (TBS, 10X) pH 8.0
Thermo Scientific Chemicals

TRIS-buffered saline (TBS, 10X) pH 8.0

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2 L
4 L
Catalog number J62780.K7
also known as J62780-K7
Price (USD)
175.65
Special offer
Online exclusive
Ends: 30-Jun-2026
206.00
Save 30.35 (15%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
175.65
Special offer
Online exclusive
Ends: 30-Jun-2026
206.00
Save 30.35 (15%)
Each
Specifications
CAS77-86-1
ChEBICHEBI:9754
Chemical Name or MaterialTRIS-buffered saline
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What are the concentrations of the components of TRIS-buffered saline (TBS, 10X) pH 8.0?

The TRIS-buffered saline (TBS, 10X) pH 8.0 contains 250 mM Tris, 27 mM KCl, and 1.37 M NaCl.