Vancomycin hydrochloride, Molecular Biology Grade
Vancomycin hydrochloride, Molecular Biology Grade
Vancomycin hydrochloride, Molecular Biology Grade
Thermo Scientific Chemicals

Vancomycin hydrochloride, Molecular Biology Grade

Vancomycin hydrochloride (vancomycin HCl, CAS # 1404-93-9) is the hydrochloride salt of the bactericidal agent, vancomycin. | CAS: 1404-93-9 | C66H76Cl3N9O24 | 1485.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62790.06
also known as J62790-06
Price (USD)
315.65
Online Exclusive
351.00
Save 35.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
315.65
Online Exclusive
351.00
Save 35.35 (10%)
Each
Chemical Identifiers
CAS1404-93-9
IUPAC Name48-({3-[(4-amino-5-hydroxy-4,6-dimethyloxan-2-yl)oxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl}oxy)-22-(carbamoylmethyl)-5,15-dichloro-2,18,32,35,37-pentahydroxy-19-[4-methyl-2-(methylamino)pentanamido]-20,23,26,42,44-pentaoxo-7,13-dioxa-21,24,27,41,43-pentaazaoctacyclo[26.14.2.2³,⁶.2¹⁴,¹⁷.1⁸,¹².1²⁹,³³.0¹⁰,²⁵.0³⁴,³⁹]pentaconta-3,5,8(48),9,11,14,16,29(45),30,32,34,36,38,46,49-pentadecaene-40-carboxylic acid hydrochloride
Molecular FormulaC66H76Cl3N9O24
InChI KeyLCTORFDMHNKUSG-UHFFFAOYNA-N
SMILESCl.CNC(CC(C)C)C(=O)NC1C(O)C2=CC(Cl)=C(OC3=CC4=CC(OC5=C(Cl)C=C(C=C5)C(O)C5NC(=O)C(NC(=O)C4NC(=O)C(CC(N)=O)NC1=O)C1=CC(=C(O)C=C1)C1=C(O)C=C(O)C=C1C(NC5=O)C(O)=O)=C3OC1OC(CO)C(O)C(O)C1OC1CC(C)(N)C(O)C(C)O1)C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan to brown
FormFree-flowing powder
Assay (HPLC)Vancomycin B: ≥85.0%
Heavy Metals≤30 ppm
Water Content (Karl Fischer Titration)≤5.0%
View more
Inhibits bacterial mucopeptide synthesis.Vancomycin hydrochloride is used as an antiviral and anti bacterial. binds to bacterial cell wall. It exhibits potent activity against Gram positive bacteria and is highly effective against MRSA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Vancomycin hydrochloride is a glycopeptide antibiotic that prevents cell wall synthesis
  • Originally obtained from the bacterial strain Streptomyces Orientalis
  • Vancomycin binds to D-alanyl-D-alanine motifs in cell wall precursors and leads to cell death
  • It is active against some penicillin-resistant bacteria

Application

  • Vancomycin hydrochloride is intended for use in the research of infections and for antibiotic resistance testing
RUO – Research Use Only

General References:

  1. Yang, Z.; Liu, J.; Gao, J.; Chen, S.; Huang, G. Chitosan coated vancomycin hydrochloride liposomes: Characterizations and evaluation. Int. J. Pharm. 2015, 495 (1), 508-515.
  2. Mori, M.; Rossi, S.; Bonferoni, M. C.; Ferrari, F.; Sandri, G.; Riva, F.; Fantec, C. D.; Perotti, C.; Caramella, C. Calcium alginate particles for the combined delivery of platelet lysate and vancomycin hydrochloride in chronic skin ulcers. Int. J. Pharm. 2014, 461 (1-2), 505-513.