Nefazodone hydrochloride, 98+%
Nefazodone hydrochloride, 98+%
Nefazodone hydrochloride, 98+%
Nefazodone hydrochloride, 98+%
Thermo Scientific Chemicals

Nefazodone hydrochloride, 98+%

CAS: 82752-99-6 | C25H33Cl2N5O2 | 506.47 g/mol
Have Questions?
Catalog NumberQuantity
J62793.03
also known as J62793-03
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62793.03
also known as J62793-03
Price (USD)
161.00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
161.00
Each
Add to cart
Specifications
CAS82752-99-6
ChEBICHEBI:7495
Chemical Name or MaterialNefazodone hydrochloride
ColorWhite
Formula Weight506.47
View more
An antidepressant acts by modifying serotonin transmission. Mixed 5-HT2A serotonin receptor antagonist/serotonin uptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An antidepressant acts by modifying serotonin transmission
  • Mixed 5-HT2A serotonin receptor antagonist/serotonin uptake inhibitor
RUO – Research Use Only