Ketanserin tartrate, 98+%
Ketanserin tartrate, 98+%
Ketanserin tartrate, 98+%
Ketanserin tartrate, 98+%
Thermo Scientific Chemicals

Ketanserin tartrate, 98+%

Selective 5-HT2A serotonin receptor antagonist | CAS: 83846-83-7 | C26H30FN3O10 | 563.54 g/mol
Have Questions?
Catalog NumberQuantity
J62798.MD
also known as J62798-MD
250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62798.MD
also known as J62798-MD
Price (USD)
412.65
Online Exclusive
458.00
Save 45.35 (10%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
412.65
Online Exclusive
458.00
Save 45.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS83846-83-7
SpecificationsSpecification SheetSpecification Sheet
Formpowder
Assay (HPLC)> 98.0%
Appearance (Color)White to off-white
Selective 5-HT2A serotonin receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only