3-Tropanylindole-3-carboxylate hydrochloride, Thermo Scientific Chemicals
3-Tropanylindole-3-carboxylate hydrochloride, Thermo Scientific Chemicals
3-Tropanylindole-3-carboxylate hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Tropanylindole-3-carboxylate hydrochloride, Thermo Scientific Chemicals

A selective 5-HT3 serotonin receptor antagonist
Have Questions?
Catalog NumberQuantity
J62807.MC
also known as J62807-MC
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62807.MC
also known as J62807-MC
Price (USD)
205.65
Special offer
Online exclusive
Ends: 30-Jun-2026
242.00
Save 36.35 (15%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
205.65
Special offer
Online exclusive
Ends: 30-Jun-2026
242.00
Save 36.35 (15%)
Each
Specifications
CAS105826-92-4
Chemical Name or Material3-Tropanylindole-3-carboxylate hydrochloride
ColorWhite
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Formula Weight320.82
View more
A selective 5-HT3 serotonin receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only