Sodium bicarbonate, 1M buffer soln., pH 9.4
Sodium bicarbonate, 1M buffer soln., pH 9.4
Sodium bicarbonate, 1M buffer soln., pH 9.4
Thermo Scientific Chemicals

Sodium bicarbonate, 1M buffer soln., pH 9.4

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J62808.AK
also known as J62808-AK
Price (USD)
89.70
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
89.70
Each
Specifications
CAS144-55-8
ChEBICHEBI:32139
Chemical Name or MaterialSodium bicarbonate buffer soln.
Health Hazard 1H319
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
View more
The bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. GL Ellman.; KD Courtney.; V Andres. A new and rapid colorimetric determination of acetylcholinesterase activity. Biochemical Pharmacology. 1961, 7, (2), 88-90.
  2. DC Rijken.; D Collen. Purification and characterization of the plasminogen activator secreted by human melanoma cells in culture. The Journal of Biological Chemistry. 1981, 256, (2), 7035-7041.