Trimyristin, 95%
Trimyristin, 95%
Trimyristin, 95%
Thermo Scientific Chemicals

Trimyristin, 95%

CAS: 555-45-3 | C45H86O6 | 723.177 g/mol
Have Questions?
Catalog NumberQuantity
J62819.14
also known as J62819-14
25 g
Catalog number J62819.14
also known as J62819-14
Price (USD)
304.65
Online Exclusive
338.00
Save 33.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
304.65
Online Exclusive
338.00
Save 33.35 (10%)
Each
Chemical Identifiers
CAS555-45-3
IUPAC Name1,3-bis(tetradecanoyloxy)propan-2-yl tetradecanoate
Molecular FormulaC45H86O6
InChI KeyDUXYWXYOBMKGIN-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay from Suppliers CofA≥94.0%
Melting Point (clear melt)55-60°C
It can be used for the preparation of solid lipid nanoparticles (SLN) composed of trimyristin (solid lipid) and poloxamer 407 (surfactant). It is also used as a synthesis reagent and as an emollient, refatting agent, skin conditioning agent, solvents and viscosity controlling agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used for the preparation of solid lipid nanoparticles (SLN) composed of trimyristin (solid lipid) and poloxamer 407 (surfactant). It is also used as a synthesis reagent and as an emollient, refatting agent, skin conditioni
RUO – Research Use Only

General References:

  1. A. van Langevelde, et al. Structure of [beta]-trimyristin and [beta]-tristearin from high-resolution X-ray powder diffraction data.Acta Cryst.,2001,B57372-377.
  2. Martins, S, et al. Improving Oral Absorption of Samon Calcitonin by Trimyristin Lipid Nanoparticles.Journal of Biomedical Nanotechnology.,2009,5(1), 76-83.