Omeprazole, 98%
Omeprazole, 98%
Omeprazole, 98%
Thermo Scientific Chemicals

Omeprazole, 98%

A proton pump inhibitor | CAS: 73590-58-6 | C17H19N3O3S | 345.417 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62860.03
also known as J62860-03
Price (USD)
65.10
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
65.10
Each
Specifications
CAS73590-58-6
ChEBICHEBI:77260
Chemical Name or MaterialOmeprazole
ColorBrown to White
Formula Weight345.42
View more
Omeprazole also acts as an aryl hydrocarbon-like inducer of cytochrome P450 secretion in human liver. This compound is an inhibitor a variety of CYP isoenzymes, including CYP2C19, CYP2C9 and CYP3A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Omeprazole also acts as an aryl hydrocarbon-like inducer of cytochrome P450 secretion in human liver. This compound is an inhibitor a variety of CYP isoenzymes, including CYP2C19, CYP2C9 and CYP3A.

Solubility
Soluble in water (0.5 mg/ml), DMSO (25 mg/ml), and ethanol (4.5 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Satoh et al. Antisecretory and anitulcer activities of a novel proton pump inhibitor AG-1749 in dogs and rats. J.Pharmacol.Exp.Ther. 1989, 248(2), 806-15.
  2. Schmarda et al. The gastric H,K-ATPase blocker lansoprazole is an inhibitor of chloride channels. Br.J.Pharmacol. 2000, 129(3), 598-604.