This product is animal origin and can only be shipped within the United States
RIPA buffer with Triton∣r X-100 (5X)
Thermo Scientific Chemicals
RIPA buffer with Triton∣r X-100 (5X)
Have Questions?
Change viewbuttonViewtableView
Quantity:
50mL
100mL
Catalog number J62885.AD
also known as J62885-AD
Price (USD)
-
Quantity:
50mL
Request bulk or custom format
Specifications
Concentration5X
FormatLiquid
Quantity50mL
Recommended StorageKeep cold
Product TypeCell Lysis Reagent
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2° to 8°C
Frequently asked questions (FAQs)
What are the concentrations of the components in RIPA buffer with Triton X-100 (5X) (Cat. No. J62885.xx)?
RIPA buffer with Triton X-100 (5X) contains 250 mM Tris-HCl (pH 7.4), 750 mM NaCl, 5% Triton X-100, 2.5% sodium deoxycholate, and 0.5% SDS.