Contents & Storage
2° to 8°C
Frequently asked questions (FAQs)
What are the concentrations of the components in RIPA buffer with Triton X-100 (5X) (Cat. No. J62885.xx)?
RIPA buffer with Triton X-100 (5X) contains 250 mM Tris-HCl (pH 7.4), 750 mM NaCl, 5% Triton X-100, 2.5% sodium deoxycholate, and 0.5% SDS.