Chelerythrine chloride, 99+%
Chelerythrine chloride, 99+%
Chelerythrine chloride, 99+%
Thermo Scientific Chemicals

Chelerythrine chloride, 99+%

CAS: 3895-92-9 | C21H18ClNO4 | 383.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
25 mg
Catalog number J62906.LB0
also known as J62906-#0
Price (USD)
130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
153.00
Save 22.35 (15%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
153.00
Save 22.35 (15%)
Each
Chemical Identifiers
CAS3895-92-9
IUPAC Name17,18-dimethoxy-21-methyl-5,7-dioxa-21-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁴,¹⁹]henicosa-1(13),2,4(8),9,11,14,16,18,20-nonaen-21-ium chloride
Molecular FormulaC21H18ClNO4
InChI KeyWEEFNMFMNMASJY-UHFFFAOYSA-M
SMILES[Cl-].COC1=CC=C2C(C=[N+](C)C3=C2C=CC2=CC4=C(OCO4)C=C32)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay (HPLC)≥99.0%
FormSolid
A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A natural benzophenanthridine alkaloid
  • Inhibits protein kinase C
  • Induces apoptosis in leukemia cells
RUO – Research Use Only