What is the maximum capacity of ethidium bromide for each bag for the Ethidium bromide de-staining bags, with activation soln.?
Each ethidium bromide de-staining bag has a maximum capacity of 5 mg of ethidium bromide.
Search
View additional product information for Ethidium bromide de-staining bags, with activation soln. - FAQs (J62931.8U)
3 product FAQs found