What are the concentrations of the components of the RBC lysis buffer for human (Cat. No. J62990.xx)?
RBC lysis buffer for human contains 150 mM ammonium chloride, 10 mM potassium bicarbonate, and 0.1 mM EDTA.
Search
View additional product information for RBC lysis buffer for human - FAQs (J62990.AK, J62990.AP)
1 product FAQs found