1-Naphthyl phosphate monosodium salt monohydrate, 97+%
1-Naphthyl phosphate monosodium salt monohydrate, 97+%
1-Naphthyl phosphate monosodium salt monohydrate, 97+%
Thermo Scientific Chemicals

1-Naphthyl phosphate monosodium salt monohydrate, 97+%

Substrate for determination of phosphatases | CAS: 81012-89-7 | C10H7O4P | 222.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J63004.06
also known as J63004-06
Price (USD)
96.60
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
96.60
Each
Chemical Identifiers
CAS81012-89-7
SpecificationsSpecification SheetSpecification Sheet
SolutionSolubility (0.5% in water): Clear solution
Solution TestSolubility (0.5% in water): Clear solution
Appearance (Color)Off white to light pink
FormSolid
Assay (Titration)≥ 97.0 %
View more
Substrate for determination of phosphatases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only