DL-Adrenaline
DL-Adrenaline
DL-Adrenaline
Thermo Scientific Chemicals

DL-Adrenaline

A hormone and neurotransmitter | CAS: 329-65-7 | C9H13NO3 | 183.207 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number J63005.03
also known as J63005-03
Price (USD)
37.50
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
37.50
Each
Specifications
CAS329-65-7
ChEBICHEBI:33568
Chemical Name or MaterialDL-Adrenaline
ColorWhite
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
View more
A hormone and neurotransmitter

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only