Phosphoenolpyruvic acid trisodium salt heptahydrate, 98%
Phosphoenolpyruvic acid trisodium salt heptahydrate, 98%
Phosphoenolpyruvic acid trisodium salt heptahydrate, 98%
Thermo Scientific Chemicals

Phosphoenolpyruvic acid trisodium salt heptahydrate, 98%

When coupled with pyruvate kinase it completes an ATP-regenerating system | CAS: 5541-93-5 | C3H2Na3O6P | 233.986 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J63013.06
also known as J63013-06
Price (USD)
800.65
Special offer
Online exclusive
Ends: 30-Jun-2026
942.00
Save 141.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
800.65
Special offer
Online exclusive
Ends: 30-Jun-2026
942.00
Save 141.35 (15%)
Each
Chemical Identifiers
CAS5541-93-5
IUPAC Nametrisodium 2-(phosphonatooxy)prop-2-enoate
Molecular FormulaC3H2Na3O6P
InChI KeyRJUFBDMHZGRGMA-UHFFFAOYSA-K
SMILES[Na+].[Na+].[Na+].[O-]C(=O)C(=C)OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Water Content33.0-37.0%
Infrared spectrumConforms
Appearance (Color)White
Assay from Supplier's CofA≥97.0%
When coupled with pyruvate kinase it completes an ATP-regenerating system

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only