How can I prepare a 1X buffer solution from the 10X Transfer or electro blotting buffer (10X), pH 8.5?
You can make a 1:10 dilution of 10X Transfer or electro blotting buffer (10X), pH 8.5 with deionized water to create a 1X Transfer or electro blotting buffer. Adjust the pH to 8.5.