Tyrphostin A9, 98+%
Tyrphostin A9, 98+%
Tyrphostin A9, 98+%
Thermo Scientific Chemicals

Tyrphostin A9, 98+%

Selective inhibitor of PDGF receptor tyrosine kinase | CAS: 10537-47-0 | C18H22N2O | 282.387 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
100 mg
Catalog number J63058.MF
also known as J63058-MF
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Add to cart
Specifications
CAS10537-47-0
ChEBICHEBI:82168
Chemical Name or MaterialTyrphostin A9
ColorYellow
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
View more
Selective inhibitor of PDGF receptor tyrosine kinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only