Nabumetone
Nabumetone
Nabumetone
Thermo Scientific Chemicals

Nabumetone

CAS: 42924-53-8 | C15H16O2 | 228.291 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63072.14
also known as J63072-14
Price (USD)
343.65
Special offer
Online exclusive
Ends: 30-Jun-2026
404.00
Save 60.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
343.65
Special offer
Online exclusive
Ends: 30-Jun-2026
404.00
Save 60.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS42924-53-8
IUPAC Name4-(6-methoxynaphthalen-2-yl)butan-2-one
Molecular FormulaC15H16O2
InChI KeyBLXXJMDCKKHMKV-UHFFFAOYSA-N
SMILESCOC1=CC=C2C=C(CCC(C)=O)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0% min.
Anti-inflammatory, anti-bacterial. A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Anti-inflammatory, anti-bacterial
  • A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase
RUO – Research Use Only