BioAcryl-P (40%, 29:1)
Thermo Scientific Chemicals

BioAcryl-P (40%, 29:1)

BioAcryl-P (Acrylamide/Bisacrylamide 29:1, CAS # 25034-58-6) is a powder-based formula used to create the gel matrix necessary for electrophoresis experiments.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J63079.AP
also known as J63079-AP
Price (USD)
125.65
Special offer
Online exclusive
Ends: 30-Jun-2026
148.00
Save 22.35 (15%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
125.65
Special offer
Online exclusive
Ends: 30-Jun-2026
148.00
Save 22.35 (15%)
Each
Specifications
Chemical Name or MaterialAcrylamide/Bisacrylamide 29:1
Concentration40% soln
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE SOLUTION
Health Hazard 1H302-H315-H317-H319-H340-H350-H361f-H372
Health Hazard 2GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
View more
Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • BioAcryl-P is a 29:1 ratio of Acrylamide/Bisacrylamide
  • Acrylamide is a crystalline amide that forms polymer structures at room temperature
  • Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation

Application

  • Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Sunyer R, Jin AJ, Nossal R, Sackett DL. Fabrication of hydrogels with steep stiffness gradients for studying cell mechanical response. PLoS One. 2012;7(10):e46107. doi: 10.1371/journal.pone.0046107. Epub 2012 Oct 4. PMID: 23056241; PMCID: PMC3464269.
  2. Diouf A, Bouchikhi B, El Bari N. A nonenzymatic electrochemical glucose sensor based on molecularly imprinted polymer and its application in measuring saliva glucose. Mater Sci Eng C Mater Biol Appl. 2019 May;98:1196-1209. doi: 10.1016/j.msec.2019.01.001. Epub 2019 Jan 11. PMID: 30813003.