Ecdysterone
Ecdysterone
Ecdysterone
Thermo Scientific Chemicals

Ecdysterone

An insect steroid hormone that induces apoptosis | CAS: 5289-74-7 | C27H44O7 | 480.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63091.MA
also known as J63091-MA
Price (USD)
225.65
Online Exclusive
251.00
Save 25.35 (10%)
Each
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)
225.65
Online Exclusive
251.00
Save 25.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS5289-74-7
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥95.0% (HPLC)
Appearance (Color)White
FormPowder
Ecdysterone is an insect steroid hormone that induces apoptosis, cell proliferation, growth by controlling gene expression involved in molting and metamorphosis. And it is also used as a ecdysteroid in both plant and animal species that controls cell death.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ecdysterone is an insect steroid hormone that induces apoptosis, cell proliferation, growth by controlling gene expression involved in molting and metamorphosis. And it is also used as a ecdysteroid in both plant and animal species that controls cell death.

Solubility
Soluble in alcohol

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Robert C. Ireland.; Edward Berger.; Karl Sirotkin.; Mary Alice Yund.; David Osterbur.; James Fristrom. Ecdysterone induces the transcription of four heat-shock genes in Drosophila S3 cells and imaginal discs ☆. Developmental Biology. 1982, 93, (2), 498-507.
  2. Herbert Oberlander. Effects of ecdysone, ecdysterone, and inokosterone on the in vitro initiation of metamorphosis of wing disks of Galleria mellonella. Journal of Insect Physiology. 1969, 15, (2), 297-304.