Esculin sesquihydrate
Esculin sesquihydrate
Esculin sesquihydrate
Thermo Scientific Chemicals

Esculin sesquihydrate

A naturally occuring antioxidant | CAS: 66778-17-4 | C30H38O21 | 734.613 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J63114.06
also known as J63114-06
Price (USD)
57.65
Special offer
Online exclusive
Ends: 30-Jun-2026
68.20
Save 10.55 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
Ends: 30-Jun-2026
68.20
Save 10.55 (15%)
Each
Chemical Identifiers
CAS66778-17-4
IUPAC Namebis(7-hydroxy-6-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one) trihydrate
Molecular FormulaC30H38O21
InChI KeyLGMHQDJZWHRZPO-UHFFFAOYNA-N
SMILESO.O.O.OCC1OC(OC2=C(O)C=C3OC(=O)C=CC3=C2)C(O)C(O)C1O.OCC1OC(OC2=C(O)C=C3OC(=O)C=CC3=C2)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content6.5-8.0%
Appearance (Color)White to off-white
Heavy Metals≤20 ppm
pH3.5-4.5 (1% in water)
Purity≥95.0% (HPLC)
View more
A naturally occuring antioxidant

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only