(+)-Dehydroabietylamine hydrochloride
(+)-Dehydroabietylamine hydrochloride
(+)-Dehydroabietylamine hydrochloride
Thermo Scientific Chemicals

(+)-Dehydroabietylamine hydrochloride

CAS: 16496-99-4 | C20H32ClN | 321.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
Catalog number J63123.MF
also known as J63123-MF
Price (USD)
564.65
Online Exclusive
627.00
Save 62.35 (10%)
Each
Quantity:
50 mg
Request bulk or custom format
Price (USD)
564.65
Online Exclusive
627.00
Save 62.35 (10%)
Each
Specifications
Beilstein3084620
CAS16496-99-4
Chemical Name or Material(+)-Dehydroabietylamine hydrochloride
Formula Weight321.9
Health Hazard 1H315-H319-H335
View more
Resolving agent for carboxylic acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only