Calcitonin, human, 96%
Thermo Scientific Chemicals

Calcitonin, human, 96%

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 mg
1 mg
Catalog number J63192.EXD
also known as J63192-!D
Price (USD)
268.65
Online Exclusive
298.00
Save 29.35 (10%)
Each
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)
268.65
Online Exclusive
298.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS21215-62-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder
Mass SpecReport Result
Purity≥95.0% (HPLC)
Decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes. A carrier peptide that can be used to internalize fusion proteins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C