Mitomycin C - FAQs
Mitomycin C - FAQs
View additional product information for Mitomycin C, Thermo Scientific Chemicals - FAQs (J63193.MA)
3 product FAQs found
How should I store Mitomycin C?
We recommend storing Mitomycin C powder protected from light, at 2-8 degrees C.
How should I store solutions of Mitomycin C in water or DMSO?
We recommend solutions of Mitomycin C in water or DMSO be stored protected from light at -20 degrees C. Solutions in water are typically stable for up to 1 week at -20 degrees C. Solutions in DMSO are typically stable for up to 1 month at -20 degrees C.
What is the solubility of Mitomycin C?
Mitomycin C is soluble at a concentration of 5 mM in water and 100 mM in DMSO.