Product Image
Thermo Scientific Chemicals
LB Agar plate with 100μg/mL Ampicillin
LB agar plate, also known as Luria Bertani medium with agar, is used to grow Escherichia coli and other bacterial cultures.
Have Questions?
Catalog NumberQuantity
J63197.EQF
also known as J63197-=F
20 Plates
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63197.EQF
also known as J63197-=F
Price (USD)
-
Quantity:
20 Plates
Request bulk or custom format
Specifications
Diameter (Metric) Outside10 cm
For Use With (Application)For growth of Escherichia coli, plasmid and other bacterial cultures
Height (Metric)15 mm
Media TypeLB Agar
Packaging TypePlate
View more
LB Agar plate with 100μg/ml ampicillin is used with recombinant strains of E.coli containing plasmids with the resistance gene to Ampicillin such as pBluescript and pUC series of plasmids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • LB agar plate is a nutrient-rich media frequently used to culture bacteria in the lab, including Escherichia coli
Application
  • LB agar plate is used to culture bacteria in microbiology research. These plates are commonly used during cloning experiments to amplify antibiotic-resistant, competent bacteria
  • The addition of agar to LB medium helps form a gel that allows bacteria to grow. The bacteria cannot digest the agar but can obtain the needed nutrients to grow from the LB medium
  • The addition of ampicillin (100 μg/mL) to these plates allows selection of the bacteria that can grow in the presence of ampicillin which is conferred by a plasmid carrying the antibiotic resistance gene
RUO – Research Use Only
General References:
  1. Park CS, Yang HJ, Kim DH, Kang DO, Kim MS, Choi NS. A screening method for β-glucan hydrolase employing Trypan Blue-coupled β-glucan agar plate and β-glucan zymography. Biotechnol Lett. 2012 Jun;34(6):1073-7. doi: 10.1007/s10529-012-0873-z. Epub 2012 Feb 15. PMID: 22350291.
  2. Rattanasuk S, Songsaeng A, Sriwarom T. Pseudomonas stutzeri CM1, Novel Thermotolerant Cellulase- Producing Bacteria Isolated from Forest Soil. Pak J Biol Sci. 2020 Jan;23(10):1345-1350. doi: 10.3923/pjbs.2020.1345.1350. PMID: 32981269.
  3. Y Doi.; D de Oliveira Garcia.; J Adams. Coproduction of novel 16S rRNA methylase RmtD and metallo-β-lactamase SPM-1 in a panresistant Pseudomonas aeruginosa isolate from Brazil. Antimicrob. Agents Chemother. 2007, 51, (3), 852-856.
  4. SA Ong.; T Peterson.; JB Neilands. Agrobactin, a siderophore from Agrobacterium tumefaciens. Journal of Biological Chemistry. 1979, 254, 1860-1865.
Contents & Storage
2°C to 8°C