Saponin permeating solution, 0.5% w/v soln. in PBS (10X) - FAQs
Saponin permeating solution, 0.5% w/v soln. in PBS (10X) - FAQs
View additional product information for Saponin permeating solution, 0.5% w/v soln. in PBS (10X), Thermo Scientific Chemicals - FAQs (J63209.AK)
1 product FAQs found
What is the concentration of Saponin in Saponin permeating solution, 0.5% w/v soln. in PBS (10X) (Cat. No. J63209.AK)?
The Saponin concentration in Saponin permeating solution, 0.5% w/v soln. in PBS (10X) (Cat. No. J63209.AK) is 0.5% w/v.