Bradykinin (1-5), Thermo Scientific Chemicals
Bradykinin (1-5), Thermo Scientific Chemicals
Bradykinin (1-5), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bradykinin (1-5), Thermo Scientific Chemicals

For cellular and molecular biology applications
Have Questions?
Catalog NumberQuantity
J63233.LB0
also known as J63233-#0
5 mg
Catalog number J63233.LB0
also known as J63233-#0
Price (USD)
62.65
Special offer
Online exclusive
Ends: 30-Jun-2026
73.10
Save 10.45 (14%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
62.65
Special offer
Online exclusive
Ends: 30-Jun-2026
73.10
Save 10.45 (14%)
Each
Specifications
For Use With (Application)Biologically active fragment of bradykinin
FormatPowder
Molecular Weight (g/mol)572.67
Product TypeBradykinin (1-5)
Purification MethodHPLC
View more
Biologically active fragment of bradykinin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C