PIPES, 0.5M buffer soln., pH 7.0
PIPES, 0.5M buffer soln., pH 7.0
PIPES, 0.5M buffer soln., pH 7.0
PIPES, 0.5M buffer soln., pH 7.0
Thermo Scientific Chemicals

PIPES, 0.5M buffer soln., pH 7.0

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J63234.AK
also known as J63234-AK
Price (USD)
136.65
Online Exclusive
152.00
Save 15.35 (10%)
Each
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)
136.65
Online Exclusive
152.00
Save 15.35 (10%)
Each
Add to cart
Specifications
CAS5625-37-6
ChEBICHEBI:44933
Chemical Name or MaterialPIPES buffer soln.
ColorColorless
IUPAC Name2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
View more
PIPES, is used as a buffering agent in biochemistry. PIPES has pKa near the physiological pH which makes it useful in cell culture work. PIPES has been documented minimizing lipid loss when buffering glutaraldehyde histology in plant and animal tissues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PIPES, is used as a buffering agent in biochemistry. PIPES has pKa near the physiological pH which makes it useful in cell culture work. PIPES has been documented minimizing lipid loss when buffering glutaraldehyde histology in plant and animal tissues.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. MJ Alowitz.; MM Scherer. Kinetics of nitrate, nitrite, and Cr (VI) reduction by iron metal. Environmental Science & Technology.2002, 36, (3), 299-306.
  2. JB Olmsted, GG Borisy. Ionic and nucleotide requirements for microtubule polymerization in vitro. Biochemistry. 1975, 14, (13), 2996-3005.