(+)-cis-Diltiazem hydrochloride
(+)-cis-Diltiazem hydrochloride
(+)-cis-Diltiazem hydrochloride
Thermo Scientific Chemicals

(+)-cis-Diltiazem hydrochloride

An L-type calcium channel blocker | CAS: 33286-22-5 | C22H27ClN2O4S | 450.978 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63245.09
also known as J63245-09
Price (USD)
181.65
Online Exclusive
202.00
Save 20.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
181.65
Online Exclusive
202.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS33286-22-5
IUPAC Namehydrogen (2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl acetate chloride
Molecular FormulaC22H27ClN2O4S
InChI KeyHDRXZJPWHTXQRI-BHDTVMLSSA-N
SMILES[H+].[Cl-].COC1=CC=C(C=C1)[C@@H]1SC2=CC=CC=C2N(CCN(C)C)C(=O)[C@@H]1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Suppliers CofA≥94.0%
An L-type calcium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only