What storage conditions should I use for Forskolin 98+%?
Forskolin 98+% should be stored in its desiccated powder form at -20°C. Solutions of forskolin can be stored at -20°C for up to 4 months.
Search
View additional product information for Forskolin, 98+% - FAQs (J63292.MF, J63292.MA)
2 product FAQs found