Geldanamycin, 98+%
Geldanamycin, 98+%
Geldanamycin, 98+%
Thermo Scientific Chemicals

Geldanamycin, 98+%

Benzoquinoid antibiotic that inhibits pp60src tyrosine kinase | CAS: 30562-34-6 | C29H40N2O9 | 560.644 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63397.MB
also known as J63397-MB
Price (USD)
181.65
Special offer
Online exclusive
Ends: 30-Jun-2026
214.00
Save 32.35 (15%)
Each
Quantity:
25 mg
Request bulk or custom format
Price (USD)
181.65
Special offer
Online exclusive
Ends: 30-Jun-2026
214.00
Save 32.35 (15%)
Each
Chemical Identifiers
CAS30562-34-6
IUPAC Name13-hydroxy-8,14,19-trimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate
Molecular FormulaC29H40N2O9
InChI KeyQTQAWLPCGQOSGP-UHFFFAOYNA-N
SMILESCOC1CC(C)CC2=C(OC)C(=O)C=C(NC(=O)C(C)=CC=CC(OC)C(OC(N)=O)C(C)=CC(C)C1O)C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA>98.0% (HPLC)
Appearance (Color)Yellow
FormPowder or crystals or crystalline powder
Geldanamycin binds to and inhibits the cytosolic chaperone functions of heat shock protein 90 (HSP90). HSP90 maintains the stability and functional shape of many oncogenic signaling proteins. It interferes with conformational maturation of proteins and the cellular stress response. In addition, it is a potent inhibitor of the nuclear hormone receptor family.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Geldanamycin binds to and inhibits the cytosolic chaperone functions of heat shock protein 90 (HSP90). HSP90 maintains the stability and functional shape of many oncogenic signaling proteins. It interferes with conformational maturation of proteins and the cellular stress response. In addition, it is a potent inhibitor of the nuclear hormone receptor family.

Solubility
Soluble in dimethyl sulfoxide at 100 mg/mL.

Notes
Store in a cool dry place. Keep away from oxidizing agents. Protect from heat.
RUO – Research Use Only

General References:

  1. S. Mark Roe.; Chrisostomos Prodromou.; Ronan O'Brien.; John E. Ladbury.; Peter W. Piper.; Laurence H. Pearl. Structural Basis for Inhibition of the Hsp90 Molecular Chaperone by the Antitumor Antibiotics Radicicol and Geldanamycin.J. Med. Chem. 1999, 42 (2),260-266 .