Oxytocin, 96%
Oxytocin, 96%
Oxytocin, 96%
Thermo Scientific Chemicals

Oxytocin, 96%

CAS: 50-56-6 | C45H67F3N12O14S2 | 1121.216 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63421.LB0
also known as J63421-#0
Price (USD)
216.65
Special offer
Online exclusive
Ends: 28-Dec-2026
255.00
Save 38.35
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
216.65
Special offer
Online exclusive
Ends: 28-Dec-2026
255.00
Save 38.35
Each
Chemical Identifiers
CAS50-56-6
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Assay (HPLC)>95.0%
CommentPeptide Content: 60 to 100%
GC/MS AnalysisElectrospray positive analysis shows mass ion corresponding to [M+H]⁺ and [M+H]²⁺
Solution TestSoluble in water
Oxytocin is used as a peptide hormone that acts predominantly as a neurotransmitter; stimulates uterine contraction and lactation, increases Na+ excretion; stimulates myometrial GTPase and phospholipase C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxytocin is used as a peptide hormone that acts predominantly as a neurotransmitter; stimulates uterine contraction and lactation, increases Na+ excretion; stimulates myometrial GTPase and phospholipase C.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Inga D Neumann.; Simone A Kromer.; Nicola Toschi.; Karl Ebner. Brain oxytocin inhibits the (re)activity of the hypothalamo-pituitary-adrenal axis in male rats: involvement of hypothalamic and limbic brain regions.Regulatory Peptides.2000, (1-2), 31-38.
  2. Roxanne Deslauriers.; Ian C. P. Smith.; Roderich Walter. Conformation flexibility of the neurohypophyseal hormones oxytocin and lysine-vasopressin. Carbon-13 spin-lattice relaxation study of backbone and side chains.J. Am. Chem. Soc.1974, (7), 2289-2291.