Substance P
Thermo Scientific Chemicals

Substance P

For cellular and molecular biology applications | 1347.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Catalog number J63489.LB0
also known as J63489-#0
Price (USD)
180.00
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
180.00
Each
Chemical Identifiers
CAS33507-63-0
IUPAC Name(2S)-2-{[(2S)-1-[(2S)-6-amino-2-{[(2S)-1-[(2S)-2-amino-5-[(diaminomethylidene)amino]pentanoyl]pyrrolidin-2-yl]formamido}hexanoyl]pyrrolidin-2-yl]formamido}-N-[(1S)-3-carbamoyl-1-{[(1S)-1-{[(1S)-1-[({[(1S)-1-{[(1S)-1-carbamoyl-3-(methylsulfanyl)propyl]carbamoyl}-3-methylbutyl]carbamoyl}methyl)carbamoyl]-2-phenylethyl]carbamoyl}-2-phenylethyl]carbamoyl}propyl]pentanediamide
Molecular FormulaC63H98N18O13S
InChI KeyADNPLDHMAVUMIW-CUZNLEPHSA-N
SMILESCSCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H]1CCCN1C(=O)[C@H](CCCCN)NC(=O)[C@@H]1CCCN1C(=O)[C@@H](N)CCCN=C(N)N)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLyophilized Powder
Assay from Supplier's CofAPeptide Content: Report Value
Assay from Supplier's CofAPeptide Purity: Report Value
Mass SpecExhibits correct MW
Appearance (Color)White
Substance P is used as a neurokinin NK-1 receptor agonist, neurokinin undecapeptide neurotransmitter and neuromodulator, also used in cancer cell proliferation and that inhibition of this component can induce apoptosis in cancer cells and decrease the steady state of Her2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only