Calcitonin, Salmon
Thermo Scientific Chemicals

Calcitonin, Salmon

 
Have Questions?
Catalog NumberQuantity
J63531.EXD
also known as J63531-!D
0.5 mg
Catalog number J63531.EXD
also known as J63531-!D
Price (USD)
146.65
Online Exclusive
163.00
Save 16.35 (10%)
Each
Quantity:
0.5 mg
Request bulk or custom format
Price (USD)
146.65
Online Exclusive
163.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS47931-85-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
AssayPeptide purity > 95%
CommentCounter ion: acetate
It shows hypocalcaemic and hypophosphatemic action, with amino-acid structure differing amongst mammalian species. Salmon calcitonin (salcatonin is the synthetic analogue) shows a markedly different sequence to that of the higher vertebrates as well as more potent activity. It decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only