Tacrolimus, 99+% - FAQs
Tacrolimus, 99+% - FAQs
View additional product information for Tacrolimus, 99+% - FAQs (J63571.MC, J63571.ME, J63571.MF)
2 product FAQs found
At what temperature should I store Tacrolimus, 99+%?
Tacrolimus should be stored in the freezer at -20 degrees Celsius.
What is the solubility of Tacrolimus, 99+%?
Tacrolimus may be dissolved in DMSO (50 mg/mL) or ethanol (50 mg/mL). This compound is very poorly soluble in water.