N-Octanoyl-N-methyl-D-glucamine
N-Octanoyl-N-methyl-D-glucamine
N-Octanoyl-N-methyl-D-glucamine
Thermo Scientific Chemicals

N-Octanoyl-N-methyl-D-glucamine

Non-ionic detergent for solubilizing membrane proteins | CAS: 85316-98-9 | C15H31NO6 | 321.414 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J63574.03
also known as J63574-03
Price (USD)
85.90
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
85.90
Each
Chemical Identifiers
CAS85316-98-9
IUPAC NameN-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]octanamide
Molecular FormulaC15H31NO6
InChI KeySBWGZAXBCCNRTM-CTHBEMJXSA-N
SMILESCCCCCCCC(=O)N(C)C[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow
FormPowder
Assay≥97.0%
N-Octanoyl-N-methylglucamine has been used in a study to assess disordered the N-terminus of PKA. It has also been used in a study to investigate the usefulness of sugar surfactants as solubilizing agents in parenteral formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Octanoyl-N-methylglucamine has been used in a study to assess disordered the N-terminus of PKA. It has also been used in a study to investigate the usefulness of sugar surfactants as solubilizing agents in parenteral formulations.

Solubility
Soluble in water (10%).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. S.Y. Huang; H.L. Chang; M. Goto. Preparation of Surfactant-Coated Lipase for the Esterification of Geraniol and Acetic Acid in Organic Solvents. Enzyme and Microbial Technology. 1998, 22 (7), 552-557.
  2. Kathryn Bracken; Robert A. Moss and Kaliappa G. Ragunathan. Remarkably Rapid Cleavage of a Model Phosphodiester by Complexed Ceric Ions in Aqueous Micellar Solutions. J. Am. Chem. Soc. 1997, 119 (39), 9323-9324.