D-erythro-Sphingosine, 99+%
D-erythro-Sphingosine, 99+%
D-erythro-Sphingosine, 99+%
Thermo Scientific Chemicals

D-erythro-Sphingosine, 99+%

CAS: 123-78-4 | C18H37NO2 | 299.499 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
25 mg
Catalog number J63584.MA
also known as J63584-MA
Price (USD)
109.65
Online exclusive
122.00
Save 12.35
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
109.65
Online exclusive
122.00
Save 12.35
Each
Specifications
Beilstein1727294
CAS123-78-4
ChEBICHEBI:16393
Chemical Name or MaterialD-erythro-Sphingosine
ColorWhite
View more
D-erythro-Sphingosine is used to inhibits protein kinase C and calmodulin-dependent enzymes, induces apoptosis. And also used to constituent of cell membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-erythro-Sphingosine is used to inhibits protein kinase C and calmodulin-dependent enzymes, induces apoptosis. And also used to constituent of cell membranes.

Solubility
Soluble in ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Hao Yang.; Lanny S. Liebeskind. A Concise and Scalable Synthesis of High Enantiopurity (-)-d-erythro-Sphingosine Using Peptidyl Thiol Ester-Boronic Acid Cross-Coupling.Org. Lett.2007, (16), 2993-2995.
  2. Peter Somfai.; Roger Olsson. An alternative synthesis of d-erythro-sphingosine and l-lyxo-phytosphingosineg.Tetrahedron Letters.1993, (30), 6645-6650.