Sodium borate, 0.5M buffer soln., pH 9.0
Sodium borate, 0.5M buffer soln., pH 9.0
Sodium borate, 0.5M buffer soln., pH 9.0
Thermo Scientific Chemicals

Sodium borate, 0.5M buffer soln., pH 9.0

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J63637.AK
also known as J63637-AK
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
94.00
Save 14.35 (15%)
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 30-Jun-2026
94.00
Save 14.35 (15%)
Each
Specifications
CAS1330-43-4
Chemical Name or MaterialSodium borate buffer soln.
ColorColorless
Health Hazard 1H319-H360FD
Health Hazard 2GHS H Statement
H360
May damage fertility or the unborn child.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium borate is used as anhydrous borax for proteomics research, used in the manufacturing of glazes and enamels, a component in tanning agents; curing and preserving skins, a agent used in cleaning compounds, used to artificially age wood, as a preservative, either alone or with other antiseptics against wood fungus, used in fireproofing fabrics and wood, used as antiseptic, used in detergents, an astringent for mucous membranes.

Solubility
Fully miscible.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only